missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Resistin / ADSF / RETN Protein
Recombinant Protein
Brand: enQuireBio™ QP5236-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001180303.1 | |
Resistin / ADSF / RETN Protein | |
Research Use Only | |
RETN | |
Recombinant Protein | |
E. coli | |
MSSKTLCSME EAINERIQEV AGSLIFRAIS SIGLECQSVT SRGDLATCPR GFAVTGCTCG SACGSWDVRA ETTCHCQCAG MDWTGARCCR VQP | |
Greater than 95% as determined by SDS-PAGE. |
56729 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human Resistin / ADSF / RETN induces ERK phosphorylation and activity (pERK) in SH-SY5Y cells via western blot (significanty different from control at Human Resistin / ADSF / RETN concentrations greater than 100 ng/ml). Activity was further confirmed by measuring IL-8 production in human PBMCs (typical ED50 is less than 500 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |