missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Resistin / ADSF / RETN Protein
Recombinant Protein
£80.80 - £5400.00
Specifications
Accession Number | NP_001180303.1 |
---|---|
Gene ID (Entrez) | 56729 |
Name | Resistin / ADSF / RETN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945759
|
enQuireBio™
QP5236-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925759
|
enQuireBio™
QP5236-25UG |
25 μg |
£191.00
25µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905759
|
enQuireBio™
QP5236-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935759
|
enQuireBio™
QP5236-500UG |
500 μg |
£3285.00
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915759
|
enQuireBio™
QP5236-1MG |
1 mg |
£5400.00
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001180303.1 | |
Resistin / ADSF / RETN Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human Resistin / ADSF / RETN induces ERK phosphorylation and activity (pERK) in SH-SY5Y cells via western blot (significanty different from control at Human Resistin / ADSF / RETN concentrations greater than 100 ng/ml). Activity was further confirmed by measuring IL-8 production in human PBMCs (typical ED50 is less than 500 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
56729 | |
Research Use Only | |
RETN | |
Recombinant Protein | |
E. coli | |
MSSKTLCSME EAINERIQEV AGSLIFRAIS SIGLECQSVT SRGDLATCPR GFAVTGCTCG SACGSWDVRA ETTCHCQCAG MDWTGARCCR VQP | |
Greater than 95% as determined by SDS-PAGE. |