missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulatory protein E2 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulatory protein E2, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Name | Regulatory protein E2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | E2 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008500
|
enQuireBio™
QP7368-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048500
|
enQuireBio™
QP7368-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018500
|
enQuireBio™
QP7368-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038500
|
enQuireBio™
QP7368-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058500
|
enQuireBio™
QP7368-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028500
|
enQuireBio™
QP7368-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Regulatory protein E2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
E2 | |
Recombinant Protein | |
Yeast | |
MQTPKETLSE RLSCVQDKII DHYENDSKDI DSQIQYWQLI RWENAIFFAA REHGIQTLNH QVVPAYNISK SKAHKAIELQ MALQGLAQSA YKTEDWTLQD TCEELWNTEP THCFKKGGQT VQVYFDGNKD NCMTYVAWDS VYYMTDAGTW DKTATCVSHR GLYYVKEGYN TFYIEFKSEC EKYGNTGTWE VHFGNNVIDC NDSMCSTSDD TVSATQLVKQ LQHTPSPYSS TVSVGTAKTY GQTSAATRPG HCGLAEKQHC GPVNPLLGAA TPTGNNKRRK LCSGNTTPII HLKGDRNSLK CLRYRLRKHS DHYRDISSTW HWTGAGNEKT GILTVTYHSE TQRTKFLNTV AIPDSVQILV GYMTM | |
Greater than 90% as determined by SDS-PAGE. |