missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Regulator of G-protein signaling 20 Protein

A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 20, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6604-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024490

  • £1735.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001273602.1
Regulator of G-protein signaling 20 Protein
Research Use Only
RGS20
Recombinant Protein
E. coli
EPAGASSPAG RVDGGLQMGS ERMEMRKRQM PAAQDTPGAA PGQPGAGSRG SNACCFCWCC CCSCSCLTVR NQEDQRPTIA SHELRADLPT WEESPAPTLE EVNAWAQSFD KLMVTPAGRN AFREFLRTEF SEENMLFWMA CEELKKEANK NIIEEKARII YEDYISILSP KEVSLDSRVR EVINRNMVEP SQHIFDDAQL QIYTLMHRDS YPRFMNSAVY KDLLQSLSEK SIEA
Greater than 80% as determined by SDS-PAGE.
8601
1 mg
Not determined.
Full Length of BC015614
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers