missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of G-protein signaling 20 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 20, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001273602.1 |
---|---|
Gene ID (Entrez) | 8601 |
Name | Regulator of G-protein signaling 20 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004490
|
enQuireBio™
QP6604-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024490
|
enQuireBio™
QP6604-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014490
|
enQuireBio™
QP6604-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034490
|
enQuireBio™
QP6604-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044490
|
enQuireBio™
QP6604-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054490
|
enQuireBio™
QP6604-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001273602.1 | |
Regulator of G-protein signaling 20 Protein | |
Not determined. | |
Full Length of BC015614 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
8601 | |
Research Use Only | |
RGS20 | |
Recombinant Protein | |
E. coli | |
EPAGASSPAG RVDGGLQMGS ERMEMRKRQM PAAQDTPGAA PGQPGAGSRG SNACCFCWCC CCSCSCLTVR NQEDQRPTIA SHELRADLPT WEESPAPTLE EVNAWAQSFD KLMVTPAGRN AFREFLRTEF SEENMLFWMA CEELKKEANK NIIEEKARII YEDYISILSP KEVSLDSRVR EVINRNMVEP SQHIFDDAQL QIYTLMHRDS YPRFMNSAVY KDLLQSLSEK SIEA | |
Greater than 80% as determined by SDS-PAGE. |