missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-5B Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-5B, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001238965.1 |
---|---|
Gene ID (Entrez) | 5869 |
Name | Ras-related protein Rab-5B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064250
|
enQuireBio™
QP6572-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084250
|
enQuireBio™
QP6572-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074250
|
enQuireBio™
QP6572-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094250
|
enQuireBio™
QP6572-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004260
|
enQuireBio™
QP6572-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014260
|
enQuireBio™
QP6572-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001238965.1 | |
Ras-related protein Rab-5B Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5869 | |
Research Use Only | |
RAB5B | |
Recombinant Protein | |
E. coli | |
TSRSTARPNG QPQASKICQF KLVLLGESAV GKSSLVLRFV KGQFHEYQES TIGAAFLTQS VCLDDTTVKF EIWDTAGQER YHSLAPMYYR GAQAAIVVYD ITNQETFARA KTWVKELQRQ ASPSIVIALA GNKADLANKR MVEYEEAQAY ADDNSLLFME TSAKTAMNVN DLFLAIAKKL PKSEPQNLGG AAGRSRGVDL HEQSQQNKSQ CCSN | |
Greater than 80% as determined by SDS-PAGE. |