missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RAD51B Protein

A cDNA sequence encoding the RAD51B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13251-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942979

  • £99.15 / 5µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RAD51B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGSKKLK RVGLSQELCD RLSRHQILTC QDFLCLSPLE LMKVTGLSYR GVHELLCMVS RACAPKMQTA YGIKAQRSAD FSPAFLSTTL SALDEALHGG VACGSLTEIT GPPGCGKTQF CIMMSILATL PTNMGGLEGA VVYIDTESAF SAERLVEIAE SRFPRYFNTE EKLLLTSSKV HLYRELTCDE VLQRIESLEE EIISKGIKLV ILDSVASVVR KEFDAQLQGN LKERNKFLAR EASSLKYLAE EFSIPVILTN QITTHLSGAL ASQADLVSPA DDLSLSEGTS GSSCVIAALG NTWSHSVNTR LILQYLDSER RQILIAKSPL APFTSFVYTI KEEGLVLQAY GNS
Greater than 85% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RAD51B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers