missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAD51B Protein
A cDNA sequence encoding the RAD51B was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | RAD51B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942979
|
enQuireBio™
QP13251-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932979
|
enQuireBio™
QP13251-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922979
|
enQuireBio™
QP13251-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RAD51B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAD51B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGSKKLK RVGLSQELCD RLSRHQILTC QDFLCLSPLE LMKVTGLSYR GVHELLCMVS RACAPKMQTA YGIKAQRSAD FSPAFLSTTL SALDEALHGG VACGSLTEIT GPPGCGKTQF CIMMSILATL PTNMGGLEGA VVYIDTESAF SAERLVEIAE SRFPRYFNTE EKLLLTSSKV HLYRELTCDE VLQRIESLEE EIISKGIKLV ILDSVASVVR KEFDAQLQGN LKERNKFLAR EASSLKYLAE EFSIPVILTN QITTHLSGAL ASQADLVSPA DDLSLSEGTS GSSCVIAALG NTWSHSVNTR LILQYLDSER RQILIAKSPL APFTSFVYTI KEEGLVLQAY GNS | |
Greater than 85% as determined by SDS-PAGE. |