missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTGS2 / COX2 / PGHS-2 Protein
A DNA sequence encoding the Homo sapiens (Human) PTGS2 / COX2 / PGHS-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6551-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000954.1 | |
PTGS2 / COX2 / PGHS-2 Protein | |
Research Use Only | |
PTGS2 | |
Recombinant Protein | |
E. coli | |
ANPCCSHPCQ NRGVCMSVGF DQYKCDCTRT GFYGENCSTP EFLTRIKLFL KPTPNTVHYI LTHFKGFWNV VNNIPFLRNA IMSYVLTSRS HLIDSPPTYN ADYGYKSWEA FSNLSYYTRA LPPVPDDCPT PLGVKGKKQL PDSNEIVEKL LLRRKFIPDP QGSNMMFAFF AQHFTHQFFK TDHKRGPAFT NGLGHGVDLN HIYGETLARQ RKLRLFKDGK MKYQIIDGEM YPPTVKDTQA EMIYPPQVPE HLRFAVGQEV FGLVPGLMMY ATIWLREHNR VCDVLKQEHP EWGDEQLFQT SRLILIGETI KIVIEDYVQH LSGYHFKLKF DPELLFNKQF QYQNRIAAEF NTLYHWHPLL PDTFQIHDQK YNYQQFIYNN SILLEHGITQ FVESFTRQIA GRVAGGRNVP PAVQKVSQAS IDQSRQMKYQ SFNEYRKRFM LKPYESFEEL TGEKEMSAEL EALYGDIDAV ELYPALLVEK PRPDAIFGET MVEVGAPFSL KGLMGNVICS PAYWKPSTFG GEVGFQIINT ASIQSLICNN VKGCPFTSFS VPDPELIKTV TINASSSRSG LDDINPTVLL KERS | |
Greater than 90% as determined by SDS-PAGE. |
5743 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |