missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTGS2 / COX2 / PGHS-2 Protein
A DNA sequence encoding the Homo sapiens (Human) PTGS2 / COX2 / PGHS-2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_000954.1 |
---|---|
Gene ID (Entrez) | 5743 |
Name | PTGS2 / COX2 / PGHS-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16074090
|
enQuireBio™
QP6551-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094090
|
enQuireBio™
QP6551-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084090
|
enQuireBio™
QP6551-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004100
|
enQuireBio™
QP6551-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014100
|
enQuireBio™
QP6551-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024100
|
enQuireBio™
QP6551-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000954.1 | |
PTGS2 / COX2 / PGHS-2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5743 | |
Research Use Only | |
PTGS2 | |
Recombinant Protein | |
E. coli | |
ANPCCSHPCQ NRGVCMSVGF DQYKCDCTRT GFYGENCSTP EFLTRIKLFL KPTPNTVHYI LTHFKGFWNV VNNIPFLRNA IMSYVLTSRS HLIDSPPTYN ADYGYKSWEA FSNLSYYTRA LPPVPDDCPT PLGVKGKKQL PDSNEIVEKL LLRRKFIPDP QGSNMMFAFF AQHFTHQFFK TDHKRGPAFT NGLGHGVDLN HIYGETLARQ RKLRLFKDGK MKYQIIDGEM YPPTVKDTQA EMIYPPQVPE HLRFAVGQEV FGLVPGLMMY ATIWLREHNR VCDVLKQEHP EWGDEQLFQT SRLILIGETI KIVIEDYVQH LSGYHFKLKF DPELLFNKQF QYQNRIAAEF NTLYHWHPLL PDTFQIHDQK YNYQQFIYNN SILLEHGITQ FVESFTRQIA GRVAGGRNVP PAVQKVSQAS IDQSRQMKYQ SFNEYRKRFM LKPYESFEEL TGEKEMSAEL EALYGDIDAV ELYPALLVEK PRPDAIFGET MVEVGAPFSL KGLMGNVICS PAYWKPSTFG GEVGFQIINT ASIQSLICNN VKGCPFTSFS VPDPELIKTV TINASSSRSG LDDINPTVLL KERS | |
Greater than 90% as determined by SDS-PAGE. |