missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein phosphatase 1 regulatory subunit 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1 regulatory subunit 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8457-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_068778.1 | |
Protein phosphatase 1 regulatory subunit 11 Protein | |
Research Use Only | |
HCGV | |
Recombinant Protein | |
E. coli | |
MAEAGAGLSE TVTETTVTVT TEPENRSLTI KLRKRKPEKK VEWTSDTVDN EHMGRRSSKC CCIYEKPRAF GESSTESDEE EEEGCGHTHC VRGHRKGRRR ATLGPTPTTP PQPPDPSQPP PGPMQH | |
Greater than 90% as determined by SDS-PAGE. |
6992 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |