missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein phosphatase 1 regulatory subunit 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1 regulatory subunit 11, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_068778.1 |
---|---|
Gene ID (Entrez) | 6992 |
Name | Protein phosphatase 1 regulatory subunit 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004731
|
enQuireBio™
QP8457-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024731
|
enQuireBio™
QP8457-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014731
|
enQuireBio™
QP8457-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034731
|
enQuireBio™
QP8457-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044731
|
enQuireBio™
QP8457-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054731
|
enQuireBio™
QP8457-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_068778.1 | |
Protein phosphatase 1 regulatory subunit 11 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6992 | |
Research Use Only | |
HCGV | |
Recombinant Protein | |
E. coli | |
MAEAGAGLSE TVTETTVTVT TEPENRSLTI KLRKRKPEKK VEWTSDTVDN EHMGRRSSKC CCIYEKPRAF GESSTESDEE EEEGCGHTHC VRGHRKGRRR ATLGPTPTTP PQPPDPSQPP PGPMQH | |
Greater than 90% as determined by SDS-PAGE. |