missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pregnancy zone Protein
A DNA sequence encoding the Homo sapiens (Human) Pregnancy zone protein, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Name | Pregnancy zone Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | PZP |
Protein Length | Partial (see sequence information for more details). of Isoform 2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094190
|
enQuireBio™
QP6563-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034200
|
enQuireBio™
QP6563-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004200
|
enQuireBio™
QP6563-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024200
|
enQuireBio™
QP6563-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044200
|
enQuireBio™
QP6563-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014200
|
enQuireBio™
QP6563-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Pregnancy zone Protein | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
PZP | |
Recombinant Protein | |
Yeast | |
MKPEAELSVS SVYNLLTVKD LTNFPDNVDQ QEEEQGHCPR PFFIHNGAIY VPLSSNEADI YSFLKGMGLK VFTNSKIRKP KSCSVIPSVS AGAVGQGYYG AGLGVVERPY VPQLGTYNVI PLNNEQSSGP VPETVRSYFP ETWIWELVAV NSSGVAEVGV TVPDTITEWK AGAFCLSEDA GLGISSTASL RAFQPFFVEL TMPYSVIRGE VFTLKATVLN YLPKCIRAEG IEQEKTFSSM TCASGANVSE QLSLKLPSNV VKESARASFS VLGDILGSAM QNIQNLLQMP YGCGEQNMVL FAPNIYVLNY LNETQQLTQE IKAKAVGYLI TGYQRQLNYK HQDGSYSTFG | |
Greater than 90% as determined by SDS-PAGE. |