missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPIH Protein
A cDNA sequence encoding the PPIH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10830-5ug
Additional Details : Weight : 0.01000kg
Specifications
10465 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 220 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
Untagged | |
1 mg/ml solution containing 1x PBS pH 7.4 10% glycerol. |
PPIH Protein | |
Research Use Only | |
PPIH | |
Recombinant Protein | |
E. coli | |
MAVANSSPVN PVVFFDVSIG GQEVGRMKIE LFADVVPKTA ENFRQFCTGEFRKDGVPIGY KGSTFHRVIK DFMIQGGDFV NGDGTGVASI YRGPFADENF KLRHSAPGLL SMANSGPSTN GCQFFITCSK CDWLDGKHVV FGKIIDGLLV MRKIENVPTG PNNKPKLPVV ISQCGEM | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |