missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPIH Protein

A cDNA sequence encoding the PPIH was constructed and used to recombinantly synthesize the protein.

£99.15 - £3965.00

Specifications

Gene ID (Entrez) 10465
Name PPIH Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol PPIH
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15965298
View Documents Promotion Details
enQuireBio™
QP10830-5UG
5 μg
£99.15
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15955298
View Documents Promotion Details
enQuireBio™
QP10830-20UG
20 μg
£155.00
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15945298
View Documents Promotion Details
enQuireBio™
QP10830-1MG
1 mg
£3965.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

10465
Research Use Only
PPIH
Recombinant Protein
E. coli
MAVANSSPVN PVVFFDVSIG GQEVGRMKIE LFADVVPKTA ENFRQFCTGEFRKDGVPIGY KGSTFHRVIK DFMIQGGDFV NGDGTGVASI YRGPFADENF KLRHSAPGLL SMANSGPSTN GCQFFITCSK CDWLDGKHVV FGKIIDGLLV MRKIENVPTG PNNKPKLPVV ISQCGEM
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
PPIH Protein
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 220 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Human
Untagged
1 mg/ml solution containing 1x PBS pH 7.4 10% glycerol.
Videos
SDS
Documents

Documents

Product Certifications