missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPIH Protein

A cDNA sequence encoding the PPIH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10830-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945298

  • £3965.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

10465
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 220 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Human
Untagged
1 mg/ml solution containing 1x PBS pH 7.4 10% glycerol.
PPIH Protein
Research Use Only
PPIH
Recombinant Protein
E. coli
MAVANSSPVN PVVFFDVSIG GQEVGRMKIE LFADVVPKTA ENFRQFCTGEFRKDGVPIGY KGSTFHRVIK DFMIQGGDFV NGDGTGVASI YRGPFADENF KLRHSAPGLL SMANSGPSTN GCQFFITCSK CDWLDGKHVV FGKIIDGLLV MRKIENVPTG PNNKPKLPVV ISQCGEM
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers