missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Pleiotrophin/PTN Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£255.93 - £680.33
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
---|---|
Gene ID (Entrez) | 5764 |
Molecular Weight (g/mol) | 15.3 kDa |
Name | Pleiotrophin/PTN |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Pleiotrophin/PTN protein Gly33-Asp168 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18344957
|
R&D Systems™
252-PL-250 |
£680.33
250µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18137380
|
R&D Systems™
252-PL-050 |
50μg |
£255.93
50µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human Pleiotrophin/PTN Protein is derived from Sf 21 (baculovirus). The Recombinant Human Pleiotrophin/PTN Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
15.3 kDa | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Pleiotrophin/PTN protein Gly33-Asp168 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
5764 | |
Pleiotrophin/PTN | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |