missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Pleiotrophin/PTN Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 252-PL-050

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18137380

  • £255.93 / 50µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Pleiotrophin/PTN Protein is derived from Sf 21 (baculovirus). The Recombinant Human Pleiotrophin/PTN Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
15.3 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
5764
Pleiotrophin/PTN
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Pleiotrophin/PTN protein Gly33-Asp168
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers