missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PIM2 Protein

A cDNA sequence encoding the PIM2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13055-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942369

  • £5730.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PIM2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLTKPLQ GPPAPPGTPT PPPGGKDREA FEAEYRLGPL LGKGGFGTVF AGHRLTDRLQ VAIKVIPRNR VLGWSPLSDS VTCPLEVALL WKVGAGGGHP GVIRLLDWFE TQEGFMLVLE RPLPAQDLFD YITEKGPLGE GPSRCFFGQV VAAIQHCHSR GVVHRDIKDE NILIDLRRGC AKLIDFGSGA LLHDEPYTDF DGTRVYSPPE WISRHQYHAL PATVWSLGIL LYDMVCGDIP FERDQEILEA ELHFPAHVSP DCCALIRRCL APKPSSRPSL EEILLDPWMQ TPAEDVPLNP SKGGPAPLAW SLLP
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PIM2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers