missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIM2 Protein
A cDNA sequence encoding the PIM2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | PIM2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952369
|
enQuireBio™
QP13055-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932369
|
enQuireBio™
QP13055-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942369
|
enQuireBio™
QP13055-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PIM2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PIM2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLTKPLQ GPPAPPGTPT PPPGGKDREA FEAEYRLGPL LGKGGFGTVF AGHRLTDRLQ VAIKVIPRNR VLGWSPLSDS VTCPLEVALL WKVGAGGGHP GVIRLLDWFE TQEGFMLVLE RPLPAQDLFD YITEKGPLGE GPSRCFFGQV VAAIQHCHSR GVVHRDIKDE NILIDLRRGC AKLIDFGSGA LLHDEPYTDF DGTRVYSPPE WISRHQYHAL PATVWSLGIL LYDMVCGDIP FERDQEILEA ELHFPAHVSP DCCALIRRCL APKPSSRPSL EEILLDPWMQ TPAEDVPLNP SKGGPAPLAW SLLP | |
Greater than 85.0% as determined by SDS-PAGE. |