missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PHOSPHO1 Protein
A DNA sequence encoding the Homo sapiens (Human) PHOSPHO1, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001137276.1 |
---|---|
Gene ID (Entrez) | 162466 |
Name | PHOSPHO1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951419
|
enQuireBio™
QP1275-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15971419
|
enQuireBio™
QP1275-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961419
|
enQuireBio™
QP1275-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15981419
|
enQuireBio™
QP1275-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15991419
|
enQuireBio™
QP1275-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15901429
|
enQuireBio™
QP1275-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001137276.1 | |
PHOSPHO1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
162466 | |
Research Use Only | |
PHOSPHO1 | |
Recombinant Protein | |
E. coli | |
MSGCFPVSGL RCLSRDGRMA AQGAPRFLLT FDFDETIVDE NSDDSIVRAA PGQRLPESLR ATYREGFYNE YMQRVFKYLG EQGVRPRDLS AIYEAIPLSP GMSDLLQFVA KQGACFEVIL ISDANTFGVE SSLRAAGHHS LFRRILSNPS GPDARGLLAL RPFHTHSCAR CPANMCKHKV LSDYLRERAH DGVHFERLFY VGDGANDFCP MGLLAGGDVA FPRRGYPMHR LIQEAQKAEP SSFRASVVPW ETAADVRLHL QQVLKSC | |
Greater than 90% as determined by SDS-PAGE. |