missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PHOSPHO1 Protein
A DNA sequence encoding the Homo sapiens (Human) PHOSPHO1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1275-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137276.1 | |
PHOSPHO1 Protein | |
Research Use Only | |
PHOSPHO1 | |
Recombinant Protein | |
E. coli | |
MSGCFPVSGL RCLSRDGRMA AQGAPRFLLT FDFDETIVDE NSDDSIVRAA PGQRLPESLR ATYREGFYNE YMQRVFKYLG EQGVRPRDLS AIYEAIPLSP GMSDLLQFVA KQGACFEVIL ISDANTFGVE SSLRAAGHHS LFRRILSNPS GPDARGLLAL RPFHTHSCAR CPANMCKHKV LSDYLRERAH DGVHFERLFY VGDGANDFCP MGLLAGGDVA FPRRGYPMHR LIQEAQKAEP SSFRASVVPW ETAADVRLHL QQVLKSC | |
Greater than 90% as determined by SDS-PAGE. |
162466 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |