missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDK / PDZ binding kinase / TOPK Protein
A DNA sequence encoding the Homo sapiens (Human) PDK / PDZ binding kinase / TOPK, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7845-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265874.1 | |
PDK / PDZ binding kinase / TOPK Protein | |
Research Use Only | |
PBK | |
Recombinant Protein | |
E. coli | |
MEGISNFKTP SKLSEKKKSV LCSTPTINIP ASPFMQKLGF GTGVNVYLMK RSPRGLSHSP WAVKKINPIC NDHYRSVYQK RLMDEAKILK SLHHPNIVGY RAFTEANDGS LCLAMEYGGE KSLNDLIEER YKASQDPFPA AIILKVALNM ARGLKYLHQE KKLLHGDIKS SNVVIKGDFE TIKICDVGVS LPLDENMTVT DPEACYIGTE PWKPKEAVEE NGVITDKADI FAFGLTLWEM MTLSIPHINL SNDDDDEDKT FDESDFDDEA YYAALGTRPP INMEELDESY QKVIELFSVC TNEDPKDRPS AAHIVEALET DV | |
Greater than 80% as determined by SDS-PAGE. |
55872 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |