missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDK / PDZ binding kinase / TOPK Protein
A DNA sequence encoding the Homo sapiens (Human) PDK / PDZ binding kinase / TOPK, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001265874.1 |
---|---|
Gene ID (Entrez) | 55872 |
Name | PDK / PDZ binding kinase / TOPK Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031131
|
enQuireBio™
QP7845-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051131
|
enQuireBio™
QP7845-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041131
|
enQuireBio™
QP7845-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061131
|
enQuireBio™
QP7845-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071131
|
enQuireBio™
QP7845-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081131
|
enQuireBio™
QP7845-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001265874.1 | |
PDK / PDZ binding kinase / TOPK Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
55872 | |
Research Use Only | |
PBK | |
Recombinant Protein | |
E. coli | |
MEGISNFKTP SKLSEKKKSV LCSTPTINIP ASPFMQKLGF GTGVNVYLMK RSPRGLSHSP WAVKKINPIC NDHYRSVYQK RLMDEAKILK SLHHPNIVGY RAFTEANDGS LCLAMEYGGE KSLNDLIEER YKASQDPFPA AIILKVALNM ARGLKYLHQE KKLLHGDIKS SNVVIKGDFE TIKICDVGVS LPLDENMTVT DPEACYIGTE PWKPKEAVEE NGVITDKADI FAFGLTLWEM MTLSIPHINL SNDDDDEDKT FDESDFDDEA YYAALGTRPP INMEELDESY QKVIELFSVC TNEDPKDRPS AAHIVEALET DV | |
Greater than 80% as determined by SDS-PAGE. |