missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDK / PDZ binding kinase / TOPK Protein

A DNA sequence encoding the Homo sapiens (Human) PDK / PDZ binding kinase / TOPK, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7845-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031131

  • £384.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265874.1
PDK / PDZ binding kinase / TOPK Protein
Research Use Only
PBK
Recombinant Protein
E. coli
MEGISNFKTP SKLSEKKKSV LCSTPTINIP ASPFMQKLGF GTGVNVYLMK RSPRGLSHSP WAVKKINPIC NDHYRSVYQK RLMDEAKILK SLHHPNIVGY RAFTEANDGS LCLAMEYGGE KSLNDLIEER YKASQDPFPA AIILKVALNM ARGLKYLHQE KKLLHGDIKS SNVVIKGDFE TIKICDVGVS LPLDENMTVT DPEACYIGTE PWKPKEAVEE NGVITDKADI FAFGLTLWEM MTLSIPHINL SNDDDDEDKT FDESDFDDEA YYAALGTRPP INMEELDESY QKVIELFSVC TNEDPKDRPS AAHIVEALET DV
Greater than 80% as determined by SDS-PAGE.
55872
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers