missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human p16-INK4a Protein

A cDNA sequence encoding the p16-INK4a was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12946-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15922039

  • £155.00 / 20µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1029
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MEPAAGSSMEPSADWLATAAARGRVEEVRALLEAGALPNAPNSYGRRPIQVMMMGSARVAELLLLHGAEPNCADPATLTRPVHDAAREGFLDTLVVLHRAGARLDVRDAWGRLPVDLAEELGHRDVARYLRAAAGGTRGSNHARIDAAEGPSDIPD
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
p16-INK4a Protein
Research Use Only
CDKN2A
Human
Untagged
CDKN2A was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1x PBS pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers