missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OVCA2 Protein

A cDNA sequence encoding the OVCA2 was constructed and used to recombinantly synthesize the protein.

£98.39 - £1324.47

Specifications

Name OVCA2 Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15942019
View Documents Promotion Details
enQuireBio™
QP12940-1UG
1 μg
£98.39
1µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15962019
View Documents Promotion Details
enQuireBio™
QP12940-5UG
5 μg
£151.37
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15952019
View Documents Promotion Details
enQuireBio™
QP12940-50UG
50 μg
£1324.47
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

OVCA2 Protein
His
Human
Greater than 90% as determined by SDS-PAGE.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAQRP LRVLCLAGFR QSERGFREKT GALRKALRGR AELVCLSGPH PVPDPPGPEG ARSDFGSCPP EEQPRGWWFS EQEADVFSAL EEPAVCRGLE ESLGMVAQAL NRLGPFDGLL GFSQGAALAA LVCALGQAGD PRFPLPRFIL LVSGFCPRGI GFKESILQRP LSLPSLHVFG DTDKVIPSQE SVQLASQFPG AITLTHSGGH FIPAAAPQRQ AYLKFLDQFA E
SDS
Product Certifications