missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OVCA2 Protein

A cDNA sequence encoding the OVCA2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12940-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962019

  • £151.37 / 5µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

OVCA2 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The OVCA2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM Nacl, 1mM DTT and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAQRP LRVLCLAGFR QSERGFREKT GALRKALRGR AELVCLSGPH PVPDPPGPEG ARSDFGSCPP EEQPRGWWFS EQEADVFSAL EEPAVCRGLE ESLGMVAQAL NRLGPFDGLL GFSQGAALAA LVCALGQAGD PRFPLPRFIL LVSGFCPRGI GFKESILQRP LSLPSLHVFG DTDKVIPSQE SVQLASQFPG AITLTHSGGH FIPAAAPQRQ AYLKFLDQFA E
Videos
SDS
Documents
Certificates
Special Offers

Special Offers