missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolar GTP-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Nucleolar GTP-binding protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7561-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037417.1 | |
Nucleolar GTP-binding protein 1 | |
Research Use Only | |
GNL2 | |
Recombinant Protein | |
E. coli | |
MVKPKYKGRS TINPSKASTN PDRVQGAGGQ NMRDRATIRR LNMYRQKERR NSRGKIIKPL QYQSTVASGT VARVEPNIKW FGNTRVIKQS SLQKFQEEMD TVMKDPYKVV MKQSKLPMSL LHDRIRPHNL KVHILDTESF ETTFGPKSQR KRPNLFASDM QSLIENAEMS TESYDQGKDR DLVTEDTGVR NEAQEEIYKK GQSKRIWGEL YKVIDSSDVV VQVLDARDPM GTRSPHIETY LKKEKPWKHL IFVLNKCDLV PTWATKRWVA VLSQDYPTLA FHASLTNPFG KGAFIQLLRQ FGKLHTDKKQ ISVGFIGYPN VGKSSVINTL RSKKVCNVAP IAGETKVWQY ITLMRRIFLI DCPGVVYPSE DSETDIVLKG VVQVEKIKSP EDHIGAVLER AKPEYISKTY KIDSWENAED FLEKLAFRTG KLLKGGEPDL QTVGKMVLND WQRGRIPFFV KPPNAEPLVA PQLLPSSSLE VVPEAAQNNP GEEVTETAGE GSESIIKEET EENSHCDANT EMQQILTRVR QNFGKINVVP QFSGDDLVPV EVSDLEEELE SFSDEEEEEQ EQQRDDAEES SSEPEEENVG NDTKAVIKAL DEKIAKYQKF LDKAKAKKFS AVRISKGLSE KIFAKPEEQR KTLEEDVDDR APSKKGKKRK AQREEEQEHS NKAPRALTSK ERRRAVRQQR PKKVGVRYYE THNVKNRNRN KKKTNDSEGQ KHKRKKFRQK Q | |
Greater than 90% as determined by SDS-PAGE. |
29889 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |