missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolar GTP-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Nucleolar GTP-binding protein 2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_037417.1 |
---|---|
Gene ID (Entrez) | 29889 |
Name | Nucleolar GTP-binding protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049390
|
enQuireBio™
QP7561-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069390
|
enQuireBio™
QP7561-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059390
|
enQuireBio™
QP7561-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079390
|
enQuireBio™
QP7561-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089390
|
enQuireBio™
QP7561-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099390
|
enQuireBio™
QP7561-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_037417.1 | |
Nucleolar GTP-binding protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
29889 | |
Research Use Only | |
GNL2 | |
Recombinant Protein | |
E. coli | |
MVKPKYKGRS TINPSKASTN PDRVQGAGGQ NMRDRATIRR LNMYRQKERR NSRGKIIKPL QYQSTVASGT VARVEPNIKW FGNTRVIKQS SLQKFQEEMD TVMKDPYKVV MKQSKLPMSL LHDRIRPHNL KVHILDTESF ETTFGPKSQR KRPNLFASDM QSLIENAEMS TESYDQGKDR DLVTEDTGVR NEAQEEIYKK GQSKRIWGEL YKVIDSSDVV VQVLDARDPM GTRSPHIETY LKKEKPWKHL IFVLNKCDLV PTWATKRWVA VLSQDYPTLA FHASLTNPFG KGAFIQLLRQ FGKLHTDKKQ ISVGFIGYPN VGKSSVINTL RSKKVCNVAP IAGETKVWQY ITLMRRIFLI DCPGVVYPSE DSETDIVLKG VVQVEKIKSP EDHIGAVLER AKPEYISKTY KIDSWENAED FLEKLAFRTG KLLKGGEPDL QTVGKMVLND WQRGRIPFFV KPPNAEPLVA PQLLPSSSLE VVPEAAQNNP GEEVTETAGE GSESIIKEET EENSHCDANT EMQQILTRVR QNFGKINVVP QFSGDDLVPV EVSDLEEELE SFSDEEEEEQ EQQRDDAEES SSEPEEENVG NDTKAVIKAL DEKIAKYQKF LDKAKAKKFS AVRISKGLSE KIFAKPEEQR KTLEEDVDDR APSKKGKKRK AQREEEQEHS NKAPRALTSK ERRRAVRQQR PKKVGVRYYE THNVKNRNRN KKKTNDSEGQ KHKRKKFRQK Q | |
Greater than 90% as determined by SDS-PAGE. |