missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear inhibitor of protein phosphatase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear inhibitor of protein phosphatase 1, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_002704.1 |
---|---|
Gene ID (Entrez) | 5511 |
Name | Nuclear inhibitor of protein phosphatase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049360
|
enQuireBio™
QP7556-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069360
|
enQuireBio™
QP7556-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059360
|
enQuireBio™
QP7556-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079360
|
enQuireBio™
QP7556-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089360
|
enQuireBio™
QP7556-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099360
|
enQuireBio™
QP7556-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002704.1 | |
Nuclear inhibitor of protein phosphatase 1 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5511 | |
Research Use Only | |
PPP1R8 | |
Recombinant Protein | |
E. coli | |
MGGEDDELKG LLGLPEEETE LDNLTEFNTA HNKRISTLTI EEGNLDIQRP KRKRKNSRVT FSEDDEIINP EDVDPSVGRF RNMVQTAVVP VKKKRVEGPG SLGLEESGSR RMQNFAFSGG LYGGLPPTHS EAGSQPHGIH GTALIGGLPM PYPNLAPDVD LTPVVPSAVN MNPAPNPAVY NPEAVNEPKK KKYAKEAWPG KKPTPSLLIL ENGTHMASSD ACHECKSMIA AAG | |
Greater than 80% as determined by SDS-PAGE. |