missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear cap-binding protein subunit 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear cap-binding protein subunit 2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001036005.1 |
---|---|
Gene ID (Entrez) | 22916 |
Name | Nuclear cap-binding protein subunit 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063090
|
enQuireBio™
QP6407-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083090
|
enQuireBio™
QP6407-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073090
|
enQuireBio™
QP6407-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093090
|
enQuireBio™
QP6407-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003100
|
enQuireBio™
QP6407-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013100
|
enQuireBio™
QP6407-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001036005.1 | |
Nuclear cap-binding protein subunit 2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
22916 | |
Research Use Only | |
NCBP2 | |
Recombinant Protein | |
E. coli | |
SGGLLKALRS DSYVELSQYR DQHFRGDNEE QEKLLKKSCT LYVGNLSFYT TEEQIYELFS KSGDIKKIIM GLDKMKKTAC GFCFVEYYSR ADAENAMRYI NGTRLDDRII RTDWDAGFKE GRQYGRGRSG GQVRDEYRQD YDAGRGGYGK LAQNQ | |
Greater than 80% as determined by SDS-PAGE. |