missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neurotrypsin Protein
A DNA sequence encoding the Homo sapiens (Human) Neurotrypsin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6537-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003610.2 | |
Neurotrypsin Protein | |
Research Use Only | |
PRSS12 | |
Recombinant Protein | |
E. coli | |
IIGGKNSLRG GWPWQVSLRL KSSHGDGRLL CGATLLSSCW VLTAAHCFKR YGNSTRSYAV RVGDYHTLVP EEFEEEIGVQ QIVIHREYRP DRSDYDIALV RLQGPEEQCA RFSSHVLPAC LPLWRERPQK TASNCYITGW GDTGRAYSRT LQQAAIPLLP KRFCEERYKG RFTGRMLCAG NLHEHKRVDS CQGDSGGPLM CERPGESWVV YGVTSWGYGC GVKDSPGVYT KVSAFVPWIK SVTK | |
Greater than 90% as determined by SDS-PAGE. |
8492 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |