missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neurotrypsin Protein
A DNA sequence encoding the Homo sapiens (Human) Neurotrypsin, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_003610.2 |
---|---|
Gene ID (Entrez) | 8492 |
Name | Neurotrypsin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094010
|
enQuireBio™
QP6537-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014020
|
enQuireBio™
QP6537-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004020
|
enQuireBio™
QP6537-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024020
|
enQuireBio™
QP6537-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034020
|
enQuireBio™
QP6537-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044020
|
enQuireBio™
QP6537-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003610.2 | |
Neurotrypsin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8492 | |
Research Use Only | |
PRSS12 | |
Recombinant Protein | |
E. coli | |
IIGGKNSLRG GWPWQVSLRL KSSHGDGRLL CGATLLSSCW VLTAAHCFKR YGNSTRSYAV RVGDYHTLVP EEFEEEIGVQ QIVIHREYRP DRSDYDIALV RLQGPEEQCA RFSSHVLPAC LPLWRERPQK TASNCYITGW GDTGRAYSRT LQQAAIPLLP KRFCEERYKG RFTGRMLCAG NLHEHKRVDS CQGDSGGPLM CERPGESWVV YGVTSWGYGC GVKDSPGVYT KVSAFVPWIK SVTK | |
Greater than 90% as determined by SDS-PAGE. |