missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MYL5 Protein

A cDNA sequence encoding the MYL5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12771-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941489

  • £155.00 / 10µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MYL5 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMASRKT KKKEGGALRA QRASSNVFSN FEQTQIQEFK EAFTLMDQNR DGFIDKEDLK DTYASLGKTN VKDDELDAML KEASGPINFT MFLNLFGEKL SGTDAEETIL NAFKMLDPDG KGKINKEYIK RLLMSQADKM TAEEVDQMFQ FASIDVAGNLDYKALSYVIT HGEEKEE
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MYL5 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl,1mM DTT and 0.1mM PMSF.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers