missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYL5 Protein
A cDNA sequence encoding the MYL5 was constructed and used to recombinantly synthesize the protein.
£99.15 - £155.00
Specifications
Name | MYL5 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951489
|
enQuireBio™
QP12771-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941489
|
enQuireBio™
QP12771-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MYL5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MYL5 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl,1mM DTT and 0.1mM PMSF. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMASRKT KKKEGGALRA QRASSNVFSN FEQTQIQEFK EAFTLMDQNR DGFIDKEDLK DTYASLGKTN VKDDELDAML KEASGPINFT MFLNLFGEKL SGTDAEETIL NAFKMLDPDG KGKINKEYIK RLLMSQADKM TAEEVDQMFQ FASIDVAGNLDYKALSYVIT HGEEKEE | |
Greater than 90.0% as determined by SDS-PAGE. |