missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPL2 Protein
A cDNA sequence encoding the MRPL2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12730-5ug
Additional Details : Weight : 0.01000kg
Specifications
MRPL2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGRDHTGR IRVHGIGGGH KQRYRMIDFL RFRPEETKSG PFEEKVIQVR YDPCRSADIA LVAGGSRKRW IIATENMQAG DTILNSNHIG RMAVAAREGD AHPLGALPVG TLINNVESEP GR | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPL2 protein solution (0.25 mg/ml) containing 20mM Phosphate buffer (pH 8.0), 1mM EDTA, 50% glycerol and 2mM DTT. |