missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPL2 Protein
A cDNA sequence encoding the MRPL2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | MRPL2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971359
|
enQuireBio™
QP12730-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961359
|
enQuireBio™
QP12730-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951359
|
enQuireBio™
QP12730-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MRPL2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPL2 protein solution (0.25 mg/ml) containing 20mM Phosphate buffer (pH 8.0), 1mM EDTA, 50% glycerol and 2mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGRDHTGR IRVHGIGGGH KQRYRMIDFL RFRPEETKSG PFEEKVIQVR YDPCRSADIA LVAGGSRKRW IIATENMQAG DTILNSNHIG RMAVAAREGD AHPLGALPVG TLINNVESEP GR | |
Greater than 90.0% as determined by SDS-PAGE. |