missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MRPL2 Protein

A cDNA sequence encoding the MRPL2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12730-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951359

  • £3965.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MRPL2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSGRDHTGR IRVHGIGGGH KQRYRMIDFL RFRPEETKSG PFEEKVIQVR YDPCRSADIA LVAGGSRKRW IIATENMQAG DTILNSNHIG RMAVAAREGD AHPLGALPVG TLINNVESEP GR
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MRPL2 protein solution (0.25 mg/ml) containing 20mM Phosphate buffer (pH 8.0), 1mM EDTA, 50% glycerol and 2mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers