missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMP 8 Protein

A cDNA sequence encoding the MMP 8 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12706-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931289

  • £2975.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4317
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLTPGNPK WERTNLTYRI RNYTPQLSEA EVERAIKDAF ELWSVASPLI FTRISQGEAD INIAFYQRDH GDNSPFDGPN GILAHAFQPG QGIGGDAHFD AEETWTNTSA NYNLFLVAAH EFGHSLGLAH SSDPGALMYP NYAFRETSNY SLPQDDIDGI QAIYGLSSNP IQPTGPSTPK PCDPSLTFDA ITTLRGEILF FKDRYFWRRH PQLQRVEMNF ISLFWPSLPT GIQAAYEDFD RDLIFLFKGN QYWALSGYDI LQGYPKDISN YGFPSSVQAI DAAVFYRSKT YFFVNDQFWR YDNQRQFMEP GYPKSISGAF PGIESKVDAV FQQEHFFHVF SGPRYYAFDL IAQRVTRVAR GNKWLNCRYG
Greater than 85% as determined by SDS-PAGE.
MMP 8 Protein
Research Use Only
MMP8
Human
His
The MMP 8 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers