missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP 8 Protein
A cDNA sequence encoding the MMP 8 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 4317 |
---|---|
Name | MMP 8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | MMP8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951289
|
enQuireBio™
QP12706-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941289
|
enQuireBio™
QP12706-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931289
|
enQuireBio™
QP12706-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
4317 | |
Research Use Only | |
MMP8 | |
Human | |
His | |
The MMP 8 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
MMP 8 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLTPGNPK WERTNLTYRI RNYTPQLSEA EVERAIKDAF ELWSVASPLI FTRISQGEAD INIAFYQRDH GDNSPFDGPN GILAHAFQPG QGIGGDAHFD AEETWTNTSA NYNLFLVAAH EFGHSLGLAH SSDPGALMYP NYAFRETSNY SLPQDDIDGI QAIYGLSSNP IQPTGPSTPK PCDPSLTFDA ITTLRGEILF FKDRYFWRRH PQLQRVEMNF ISLFWPSLPT GIQAAYEDFD RDLIFLFKGN QYWALSGYDI LQGYPKDISN YGFPSSVQAI DAAVFYRSKT YFFVNDQFWR YDNQRQFMEP GYPKSISGAF PGIESKVDAV FQQEHFFHVF SGPRYYAFDL IAQRVTRVAR GNKWLNCRYG | |
Greater than 85% as determined by SDS-PAGE. |