missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MLX-interacting Protein
A DNA sequence encoding the Homo sapiens (Human) MLX-interacting protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8054-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055753.3 | |
MLX-interacting Protein | |
Research Use Only | |
MLXIP | |
Recombinant Protein | |
E. coli | |
MDEQGCEHTS RTEDPFIQPT DFGPSEPPLS VPPPISPVLP LVPPPATALN PPAPPTFHQP QKFAGVNKAP SVITHTASAT LTHDAPATTF SQSQGLVITT HHPAPSAAPC GLALSPVTRP PQPRLTFVHP KPVSLTGGRP KQPHKIVPAP KPEPVSLVLK NARIAPGEPG GETQCGAPPD PEGCFPIPKA FKLVTTTTTL VCTCMRTHIH LNETKVS | |
Greater than 90% as determined by SDS-PAGE. |
22877 | |
100 μg | |
Not determined. | |
Full Length of Isoform 5 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |