missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MLX-interacting Protein
A DNA sequence encoding the Homo sapiens (Human) MLX-interacting protein, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_055753.3 |
---|---|
Gene ID (Entrez) | 22877 |
Name | MLX-interacting Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062391
|
enQuireBio™
QP8054-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082391
|
enQuireBio™
QP8054-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072391
|
enQuireBio™
QP8054-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092391
|
enQuireBio™
QP8054-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002401
|
enQuireBio™
QP8054-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012401
|
enQuireBio™
QP8054-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055753.3 | |
MLX-interacting Protein | |
Not determined. | |
Full Length of Isoform 5 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
22877 | |
Research Use Only | |
MLXIP | |
Recombinant Protein | |
E. coli | |
MDEQGCEHTS RTEDPFIQPT DFGPSEPPLS VPPPISPVLP LVPPPATALN PPAPPTFHQP QKFAGVNKAP SVITHTASAT LTHDAPATTF SQSQGLVITT HHPAPSAAPC GLALSPVTRP PQPRLTFVHP KPVSLTGGRP KQPHKIVPAP KPEPVSLVLK NARIAPGEPG GETQCGAPPD PEGCFPIPKA FKLVTTTTTL VCTCMRTHIH LNETKVS | |
Greater than 90% as determined by SDS-PAGE. |