missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Methionine synthase reductase Protein
A DNA sequence encoding the Homo sapiens (Human) Methionine synthase reductase, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_002445.2 |
---|---|
Gene ID (Entrez) | 4552 |
Name | Methionine synthase reductase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083421
|
enQuireBio™
QP8231-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023431
|
enQuireBio™
QP8231-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093421
|
enQuireBio™
QP8231-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013431
|
enQuireBio™
QP8231-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033431
|
enQuireBio™
QP8231-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003431
|
enQuireBio™
QP8231-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002445.2 | |
Methionine synthase reductase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4552 | |
Research Use Only | |
MTRR | |
Recombinant Protein | |
Yeast | |
MGAASVRAGA RLVEVALCSF TVTCLEVMRR FLLLYATQQG QAKAIAEEIC EQAVVHGFSA DLHCISESDK YDLKTETAPL VVVVSTTGTG DPPDTARKFV KEIQNQTLPV DFFAHLRYGL LGLGDSEYTY FCNGGKIIDK RLQELGARHF YDTGHADDCV GLELVVEPWI AGLWPALRKH FRSSRGQEEI SGALPVASPA SSRTDLVKSE LLHIESQVEL LRFDDSGRKD SEVLKQNAVN SNQSNVVIED FESSLTRSVP PLSQASLNIP GLPPEYLQVH LQESLGQEES QVSVTSADPV FQVPISKAVQ LTTNDAIKTT LLVELDISNT DFSYQPGDAF SVICPNSDSE VQSLLQRLQL EDKREHCVLL KIKADTKKKG ATLPQHIPAG CSLQFIFTWC LEIRAIPKKA FLRALVDYTS DSAEKRRLQE LCSKQGAADY SRFVRDACAC LLDLLLAFPS CQPPLSLLLE HLPKLQPRPY SCASSSLFHP GKLHFVFNIV EFLSTATTEV LRKGVCTGWL ALLVASVLQP NIHASHEDSG KALAPKISIS PRTTNSFHLP DDPSIPIIMV GPGTGIAPFI GFLQHREKLQ EQHPDGNFGA MWLFFGCRHK DRDYLFRKEL RHFLKHGILT HLKVSFSRDA PVGEEEAPAK YVQDNIQLHG QQVARILLQE NGHIYVCGDA KNMAKDVHDA LVQIISKEVG VEKLEAMKTL ATLKEEKRYL QDIWS | |
Greater than 90% as determined by SDS-PAGE. |