missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MEIS3 Protein

A cDNA sequence encoding the MEIS3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12675-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961189

  • £99.15 / 5µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MEIS3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMARRYDE LPHYPGIVDG PAALASFPET VPAVPGPYGP HRPPQPLPPG LDSDGLKREK DEIYGHPLFP LLALVFEKCE LATCSPRDGA GAGLGTPPGG DVCSSDSFNE DIAAFAKQVR SERPLFSSNP ELDNLMIQAI QVLRFHLLEL EKGKMPIDLVIEDRDGGCRE DFEDYPASCP SLPDQNNMWI RDHEDSGSVH LGTPGPSSGG LASQSGDNSS DQGDGLDTSV ASPSSGGEDE DLDQERRRNK KRGIFPKVAT NIMRAWLFQH LSHPYPSEEQ KKQLAQDTGL TILQVNNWFI NARRRIVQPM IDQSNRTGQG AAFSPEGQPI GGYTETQPHVAVRPPGSVGM SLNLEGEWHY L
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MEIS3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers