missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MEIS3 Protein
A cDNA sequence encoding the MEIS3 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | MEIS3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961189
|
enQuireBio™
QP12675-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951189
|
enQuireBio™
QP12675-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941189
|
enQuireBio™
QP12675-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MEIS3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MEIS3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMARRYDE LPHYPGIVDG PAALASFPET VPAVPGPYGP HRPPQPLPPG LDSDGLKREK DEIYGHPLFP LLALVFEKCE LATCSPRDGA GAGLGTPPGG DVCSSDSFNE DIAAFAKQVR SERPLFSSNP ELDNLMIQAI QVLRFHLLEL EKGKMPIDLVIEDRDGGCRE DFEDYPASCP SLPDQNNMWI RDHEDSGSVH LGTPGPSSGG LASQSGDNSS DQGDGLDTSV ASPSSGGEDE DLDQERRRNK KRGIFPKVAT NIMRAWLFQH LSHPYPSEEQ KKQLAQDTGL TILQVNNWFI NARRRIVQPM IDQSNRTGQG AAFSPEGQPI GGYTETQPHVAVRPPGSVGM SLNLEGEWHY L | |
Greater than 85.0% as determined by SDS-PAGE. |