missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LSM12 Protein
A cDNA sequence encoding the LSM12 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 124801 |
---|---|
Name | LSM12 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | LSM12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950919
|
enQuireBio™
QP12590-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940919
|
enQuireBio™
QP12590-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930919
|
enQuireBio™
QP12590-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
124801 | |
Research Use Only | |
LSM12 | |
Human | |
His | |
LSM12 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.1M NaCl, 10% glycerol and 1mM DTT. |
LSM12 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAPPGE YFSVGSQVSC RTCQEQRLQG EVVAFDYQSK MLALKCPSSS GKPNHADILL INLQYVSEVE IINDRTETPP PLASLNVSKL ASKARTEKEE KLSQAYAISA GVSLEGQQLF QTIHKTIKDC KWQEKNIVVM EEVVITPPYQ VENCKGKEGS ALSHVRKIVE KHFRDVESQK ILQRSQAQQP QKEAALSS | |
Greater than 90% as determined by SDS-PAGE. |