missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LSM12 Protein
A cDNA sequence encoding the LSM12 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12590-100ug
Additional Details : Weight : 0.01000kg
Specifications
124801 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAPPGE YFSVGSQVSC RTCQEQRLQG EVVAFDYQSK MLALKCPSSS GKPNHADILL INLQYVSEVE IINDRTETPP PLASLNVSKL ASKARTEKEE KLSQAYAISA GVSLEGQQLF QTIHKTIKDC KWQEKNIVVM EEVVITPPYQ VENCKGKEGS ALSHVRKIVE KHFRDVESQK ILQRSQAQQP QKEAALSS | |
Greater than 90% as determined by SDS-PAGE. |
LSM12 Protein | |
Research Use Only | |
LSM12 | |
Human | |
His | |
LSM12 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.1M NaCl, 10% glycerol and 1mM DTT. |