missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KIAA0513 Protein

A cDNA sequence encoding the KIAA0513 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12485-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960569

  • £5730.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KIAA0513 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMETPEVP VGSLIDFGPE APTSSPLEAP PPVLQDGDGS LGDGASESET TESADSENDM GESPSHPSWD QDRRSSSNES FSSNQSTEST QDEETLALRD FMRGYVEKIF SGGEDLDQEE KAKFGEYCSS ENGKGREWFA RYVSAQRCNS KCVSEATFYR LVQSFAVVLF ECHQMDDFGP AKNLMTMCFT YYHIGKPQLL PPESREKPAG SIDSYLKSAN SWLAEKKDIA ERLLKNTSAR TENVKGFFGG LETKLKGPLA RRNEEDENKP QEKRPRAVTA YSPEDEKKGE KIYLYTHLKQ QPIWHTLRFW NAAFFDAVHC ERTKRSPTTR EKWCHMTQEE RDDSLRFNEN ITFGQLGTFT HNMLAFGLNK KLCNDFLKKQ AVIGNLDEEQ YKLLSDHIEQ MATE
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
KIAA0513 protein solution (0.25 mg/ml) in Phosphate buffer saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers