missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KIAA0513 Protein
A cDNA sequence encoding the KIAA0513 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | KIAA0513 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970569
|
enQuireBio™
QP12485-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950569
|
enQuireBio™
QP12485-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960569
|
enQuireBio™
QP12485-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
KIAA0513 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KIAA0513 protein solution (0.25 mg/ml) in Phosphate buffer saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMETPEVP VGSLIDFGPE APTSSPLEAP PPVLQDGDGS LGDGASESET TESADSENDM GESPSHPSWD QDRRSSSNES FSSNQSTEST QDEETLALRD FMRGYVEKIF SGGEDLDQEE KAKFGEYCSS ENGKGREWFA RYVSAQRCNS KCVSEATFYR LVQSFAVVLF ECHQMDDFGP AKNLMTMCFT YYHIGKPQLL PPESREKPAG SIDSYLKSAN SWLAEKKDIA ERLLKNTSAR TENVKGFFGG LETKLKGPLA RRNEEDENKP QEKRPRAVTA YSPEDEKKGE KIYLYTHLKQ QPIWHTLRFW NAAFFDAVHC ERTKRSPTTR EKWCHMTQEE RDDSLRFNEN ITFGQLGTFT HNMLAFGLNK KLCNDFLKKQ AVIGNLDEEQ YKLLSDHIEQ MATE | |
Greater than 85.0% as determined by SDS-PAGE. |